Learn More
YMC America, Inc ODS-AL 120A 5um 150x0.5mm 1/32
SDP
Catalog No. 5000879479
Encompass
YMC-Pack ODS-AL is a non-endcapped high carbon load monomeric C18 phase It exhibits different selectivity from conventional ODS columns due to the use of not only hydrophobic interaction but also secondary interactions caused by silanol groups YMC-Pack ODS-AL is intended for use only in those situations where residual silanol activity is beneficial to the separation
Catalog No. 50-008-79479 Supplier YMC America, Inc Supplier No. AL12S0515J0RU
YMC-Pack ODS-AL is a non-endcapped high carbon load monomeric C18 phase It exhibits different selectivity from conventional ODS columns due to the use of not only hydrophobic interaction but also secondary interactions caused by silanol groups YMC-Pack ODS-AL is intended for use only in those situations where residual silanol activity is beneficial to the separation