Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc ODS-AL 5UM 120A 20X4MM PK/5
SDP

Catalog No. 50157214
Encompass

AL12S05-0204GC: ODS-AL, 5um, 120A, 20X4mm Cartridge, pk/5

Catalog No. 50-157-214 Supplier YMC America, Inc Supplier No. AL12S050204GC
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AL12S05-0204GC: ODS-AL, 5um, 120A, 20X4mm Cartridge, pk/5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.