Learn More
YMC America, Inc ODS-AL col 120A 3um 150x3mm
SDP
Catalog No. 5000880791
Encompass_Preferred
YMC-Pack ODS-AL is a non-endcapped high carbon load monomeric C18 phase It exhibits different selectivity from conventional ODS columns due to the use of not only hydrophobic interaction but also secondary interactions caused by silanol groups YMC-Pack ODS-AL is intended for use only in those situations where residual silanol activity is beneficial to the separation
Catalog No. 50-008-80791 Supplier YMC America, Inc Supplier No. AL12S031503WT
Add to Cart
Add to Cart
YMC-Pack ODS-AL is a non-endcapped high carbon load monomeric C18 phase It exhibits different selectivity from conventional ODS columns due to the use of not only hydrophobic interaction but also secondary interactions caused by silanol groups YMC-Pack ODS-AL is intended for use only in those situations where residual silanol activity is beneficial to the separation