Learn More
YMC America, Inc ODS-AL guard 120A 5um 10x30mm
SDP
Catalog No. 5000880790
Encompass
YMC-Pack ODS-AL is a non-endcapped high carbon load monomeric C18 phase It exhibits different selectivity from conventional ODS columns due to the use of not only hydrophobic interaction but also secondary interactions caused by silanol groups YMC-Pack ODS-AL is intended for use only in those situations where residual silanol activity is beneficial to the separation
Catalog No. 50-008-80790 Supplier YMC America, Inc Supplier No. AL12S050130CCN
YMC-Pack ODS-AL is a non-endcapped high carbon load monomeric C18 phase It exhibits different selectivity from conventional ODS columns due to the use of not only hydrophobic interaction but also secondary interactions caused by silanol groups YMC-Pack ODS-AL is intended for use only in those situations where residual silanol activity is beneficial to the separation