Learn More
YMC America, Inc ODS-AM 120A 3um 100x.15mm 1/16
SDP
Catalog No. 5000881187
Encompass
YMC-Pack ODS-AM is produced with specific emphasis on lot-to-lot reproducibility of the packing material Strict manufacturing control is performed on all aspects of quality including physical properties of the silica base gel as well as surface modifying processes ensuring exceptional reproducibility
Catalog No. 50-008-81187 Supplier YMC America, Inc Supplier No. AM12S0310F5AU
YMC-Pack ODS-AM is produced with specific emphasis on lot-to-lot reproducibility of the packing material Strict manufacturing control is performed on all aspects of quality including physical properties of the silica base gel as well as surface modifying processes ensuring exceptional reproducibility