Learn More
YMC America, Inc ODS-AM 120A 5um 5x0.5mm 1/16
SDP
Catalog No. 5000879365
Encompass
YMC-Pack ODS-AM is produced with specific emphasis on lot-to-lot reproducibility of the packing material Strict manufacturing control is performed on all aspects of quality including physical properties of the silica base gel as well as surface modifying processes ensuring exceptional reproducibility
Catalog No. 50-008-79365 Supplier YMC America, Inc Supplier No. AM12S05E5J0AU
YMC-Pack ODS-AM is produced with specific emphasis on lot-to-lot reproducibility of the packing material Strict manufacturing control is performed on all aspects of quality including physical properties of the silica base gel as well as surface modifying processes ensuring exceptional reproducibility