Learn More
YMC America, Inc ODS-AM guard 120A 5um 20x3mm
SDP
Catalog No. 5000879395
Encompass
YMC-Pack ODS-AM is produced with specific emphasis on lot-to-lot reproducibility of the packing material Strict manufacturing control is performed on all aspects of quality including physical properties of the silica base gel as well as surface modifying processes ensuring exceptional reproducibility
Catalog No. 50-008-79395 Supplier YMC America, Inc Supplier No. AM12S050203GC
YMC-Pack ODS-AM is produced with specific emphasis on lot-to-lot reproducibility of the packing material Strict manufacturing control is performed on all aspects of quality including physical properties of the silica base gel as well as surface modifying processes ensuring exceptional reproducibility