Learn More
YMC America, Inc ODS-AQ 3UM 200A 50X4MM
SDP
Catalog No. 50157091
Encompass_Preferred
AQ20S03-0504WT: ODS-AQ, 3um, 200A, 50X4mm Column
Catalog No. 50-157-091 Supplier YMC America, Inc Supplier No. AQ20S030504WT
Add to Cart
Add to Cart
AQ20S03-0504WT: ODS-AQ, 3um, 200A, 50X4mm Column