Learn More
YMC America, Inc ODS-AQHG grd 120A 10um 50x30mm
SDP
Catalog No. 5000880508
Encompass
YMC-Pack ODS-AQ is a unique reversed-phase material with both a hydrophobic high carbon loading and a relatively hydrophilic surface Because of the hydrophilic surface ODS-AQ can be wetted with polar eluents It provides chromatographers reproducible retention and excellent peak shape in highly aqueous mobile phases including 100% water This is due to the C18 ligands on ODS-AQ actually being lifted from the surface by the action of the eluents penetrating the silica pores Selectivity on YMC-Pack ODS-AQ can be subtly different from that seen on conventional ODS columns Differences in selectivity are most often encountered when analyzing mixtures of relatively polar compounds Typically longer retention and greater resolution are observed with ODS-AQ Available in both 120 and 200 pore sizes YMC-Pack ODS-AQ is well-suited for analysis and purification of peptides and other biomolecules
Catalog No. 50-008-80508 Supplier YMC America, Inc Supplier No. AQG12S110530WTG
YMC-Pack ODS-AQ is a unique reversed-phase material with both a hydrophobic high carbon loading and a relatively hydrophilic surface Because of the hydrophilic surface ODS-AQ can be wetted with polar eluents It provides chromatographers reproducible retention and excellent peak shape in highly aqueous mobile phases including 100% water This is due to the C18 ligands on ODS-AQ actually being lifted from the surface by the action of the eluents penetrating the silica pores Selectivity on YMC-Pack ODS-AQ can be subtly different from that seen on conventional ODS columns Differences in selectivity are most often encountered when analyzing mixtures of relatively polar compounds Typically longer retention and greater resolution are observed with ODS-AQ Available in both 120 and 200 pore sizes YMC-Pack ODS-AQ is well-suited for analysis and purification of peptides and other biomolecules