Learn More
Cayman Chemical O-desmethyl Brinzolamide (hydrochloride), Purity: ≥98%, CAS: 2967475-56-3, 10 mg
SDP
Catalog No. 502946191
Encompass_Preferred
An active metabolite of brinzolamide; inhibits CAII and CAIV (IC50s = 0.136 and 165 nM, respectively)
Catalog No. 50-294-6191 Supplier Cayman Chemical Supplier No. 3427610 MG
An active metabolite of brinzolamide; inhibits CAII and CAIV (IC50s = 0.136 and 165 nM, respectively)