Learn More
LICOR Odyssey, Bis-Tris, 10x8, 10%, 15 wells (pack of 10)
Catalog No. 502773675
Encompass_Preferred
Odyssey Mini Gels are polyacrylamide gels designed to separate a wide range of protein sizes via PAGE or SDS-PAGE. Each box contains 10 pre-cast gels. Our gels feature larger well volumes for easier sample loading and are cast in a neutral pH buffer to increase gel stability. They also run at neutral pH, which minimizes protein modification compared to Tris-glycine gels. Odyssey Mini Gels are available in several gradients with 10, 12, or 15 numbered wells. Our gels feature a shelf life of up to 6 months at room temperature and 12 months at 2–8C.
Catalog No. 50-277-3675 Supplier LICOR Supplier No. 92721195
Odyssey Mini Gels are polyacrylamide gels designed to separate a wide range of protein sizes via PAGE or SDS-PAGE. Each box contains 10 pre-cast gels. Our gels feature larger well volumes for easier sample loading and are cast in a neutral pH buffer to increase gel stability. They also run at neutral pH, which minimizes protein modification compared to Tris-glycine gels. Odyssey Mini Gels are available in several gradients with 10, 12, or 15 numbered wells. Our gels feature a shelf life of up to 6 months at room temperature and 12 months at 2–8C.