Learn More
LICORbio Odyssey XF Imaging Trays (pk of 5 trays)
Catalog No. NC0189648
Encompass_Preferred
Imaging trays for use with the Odyssey XF and Odyssey Fc Imagers. To use, use trays logo side up, lay sample flat on tray and within the marks at each corner, place tray in instrument drawer (notches in tray will align with tabs in drawer assembly), close drawer and proceed with imaging procedure.
Catalog No. NC0189648 Supplier LICORbio Supplier No. 92640020
Imaging trays for use with the Odyssey XF and Odyssey Fc Imagers. To use, use trays logo side up, lay sample flat on tray and within the marks at each corner, place tray in instrument drawer (notches in tray will align with tabs in drawer assembly), close drawer and proceed with imaging procedure.