Learn More
LICORbio Odyssey MES SDS Running Buffer Powder (pack of 5)
Catalog No. 502773678
Encompass_Preferred
Odyssey Mini Gels are polyacrylamide gels designed to separate a wide range of protein sizes via PAGE or SDS-PAGE. Each box contains 10 pre-cast gels. Our gels feature larger well volumes for easier sample loading and are cast in a neutral pH buffer to increase gel stability. They also run at neutral pH, which minimizes protein modification compared to Tris-glycine gels. Odyssey Mini Gels are available in several gradients with 10, 12, or 15 numbered wells. Our gels feature a shelf life of up to 6 months at room temperature and 12 months at 2–8C.
Catalog No. 50-277-3678 Supplier LICORbio Supplier No. 92621173
Odyssey Mini Gels are polyacrylamide gels designed to separate a wide range of protein sizes via PAGE or SDS-PAGE. Each box contains 10 pre-cast gels. Our gels feature larger well volumes for easier sample loading and are cast in a neutral pH buffer to increase gel stability. They also run at neutral pH, which minimizes protein modification compared to Tris-glycine gels. Odyssey Mini Gels are available in several gradients with 10, 12, or 15 numbered wells. Our gels feature a shelf life of up to 6 months at room temperature and 12 months at 2–8C.