Learn More
LICORbio Odyssey Tris-MOPS-SDS Running Buffer Powder (pack of 5)
Catalog No. 502773679
Encompass_Preferred
Odyssey MES SDS Running Buffer is formulated for running Odyssey Mini Gels or similar bis-tris gels. This buffer is ideal for separating small- and mid-sized proteins during PAGE and SDS-PAGE. For separation of large- to medium-sized proteins, we recommend using Odyssey Tris-MOPS SDS Buffer instead. Each box includes 5 individual packs of powder. Reconstitute each pack with 1000 ml deionized water to make 1X running buffer. Do not use acid or base to adjust the pH.
Catalog No. 50-277-3679 Supplier LICORbio Supplier No. 92621174
Odyssey MES SDS Running Buffer is formulated for running Odyssey Mini Gels or similar bis-tris gels. This buffer is ideal for separating small- and mid-sized proteins during PAGE and SDS-PAGE. For separation of large- to medium-sized proteins, we recommend using Odyssey Tris-MOPS SDS Buffer instead. Each box includes 5 individual packs of powder. Reconstitute each pack with 1000 ml deionized water to make 1X running buffer. Do not use acid or base to adjust the pH.