Learn More
LICORbio Odyssey Pen (pk of 4 pens)
Catalog No. NC9839078
Encompass_Preferred
The Odyssey Pen is suitable for writing on membranes to be scanned on any of the Odyssey Imagers. Inks from other pens will autofluoresce in the 700 nm channel, potentially adding to background or interfering artifacts on your image. The Odyssey Pen does not.
Catalog No. NC9839078 Supplier LICORbio Supplier No. 92671804
The Odyssey Pen is suitable for writing on membranes to be scanned on any of the Odyssey Imagers. Inks from other pens will autofluoresce in the 700 nm channel, potentially adding to background or interfering artifacts on your image. The Odyssey Pen does not.