Learn More
LICORbio Odyssey One-Color Protein Molecular Weight Marker (500 L)
Catalog No. NC9978781
Encompass_Preferred
The Odyssey One-Color Protein MW Marker provides a ladder of convenient, consistent sizes for both visible viewing and 700 nm channel for near-infrared detection. It includes ten protein bands from 10 to 250 kDa and it has three reference bands at 25, 50, and 75 kDa that are three times as intense as other bands.
Catalog No. NC9978781 Supplier LICORbio Supplier No. 92840000
The Odyssey One-Color Protein MW Marker provides a ladder of convenient, consistent sizes for both visible viewing and 700 nm channel for near-infrared detection. It includes ten protein bands from 10 to 250 kDa and it has three reference bands at 25, 50, and 75 kDa that are three times as intense as other bands.