Learn More
MSC Proto Open End Wrench, 15/16 in., Angle, 1 ea
Catalog No. 501920503
Encompass
This double-ended open end wrench is designed for professional use, featuring 15° and 60° angled heads for access in confined spaces. Constructed from steel with a chrome finish, it provides durability and corrosion resistance, meeting both ANSI and Federal specifications.
  • Satin chrome finish for good grip and easy cleaning
  • Chamfered ends at 45° allow easy placement of wrench on fastener
  • Both ends are the same size for versatile use in four positions
  • Continuous arc design minimizes spreading and enhances strength
  • Thin head for tight clearances
  • Wrench heads offset at 15° and 60° to the handle
  • Meets ANSI B107.4 and Federal specifications
Catalog No. 50-192-0503 Supplier MSC Supplier No. 39974746
This double-ended open end wrench is designed for professional use, featuring 15° and 60° angled heads for access in confined spaces. Constructed from steel with a chrome finish, it provides durability and corrosion resistance, meeting both ANSI and Federal specifications.
  • Satin chrome finish for good grip and easy cleaning
  • Chamfered ends at 45° allow easy placement of wrench on fastener
  • Both ends are the same size for versatile use in four positions
  • Continuous arc design minimizes spreading and enhances strength
  • Thin head for tight clearances
  • Wrench heads offset at 15° and 60° to the handle
  • Meets ANSI B107.4 and Federal specifications