Learn More
Mopec OE100 Dual Draft Countertop Trimming Station
SDP
Catalog No. 22444240 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Trimming Station
Catalog No. Product Type
22-444-240 Trimming Station
1 options
Catalog No. 22-444-240 Supplier Mopec Supplier No. OE100
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Highly ventilated and compact counter top workstation that ensures a safe work environment. Mopec™ OE100 Dual Draft Countertop Trimming Station is a piece of aminal sciene equiptment that is designed for laboratories with both space constraints and the need for maximum ventilation.
  • Faucet fixture with swing spout
  • Paper towel dispenser
  • One large and one small organizer bin
  • Includes a 30cm adjustable shelf
  • Energy saving, long life LED light
  • GFCI outlet
  • Inch and cm ruler
  • Magnetic tool bar
  • Perforated grid plates
  • Total draft chamber
Warranty and Services
1 Year Warranty
Specifications
Product Type Trimming Station
Includes 30 cm adjustable shelf
Electrical Requirements GFCI Outlet
Ventilation Total Draft Chamber
Elevating or Fixed Height Fixed