Learn More
Sigma Aldrich Fine Chemicals Biosciences OECM 1 Human Oral Squamous
Catalog No. 502908187
Encompass
The OECM-1 human oral cavity squamous cell carcinoma cell line is a well-established model for squamous cell carcinoma. The OECM-1 cell line harbors a missense mutation in the p53 tumor suppressor displays low EGFR expression and is tumorigenic in nude mice . OECM-1 cells proliferate with a doubling time of 30-38 hours and are capable of anchorage-independent growth forming spheroid colonies . OECM-1 cells have been intensively characterized in the literature for morphology biomarker expression and drug response .SourceThe OECM-1 human oral cavity squamous cell carcinoma cell line was derived from surgical resection of a primary tumor of a Taiwanese male patient .
Catalog No. 50-290-8187 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SCC180
The OECM-1 human oral cavity squamous cell carcinoma cell line is a well-established model for squamous cell carcinoma. The OECM-1 cell line harbors a missense mutation in the p53 tumor suppressor displays low EGFR expression and is tumorigenic in nude mice . OECM-1 cells proliferate with a doubling time of 30-38 hours and are capable of anchorage-independent growth forming spheroid colonies . OECM-1 cells have been intensively characterized in the literature for morphology biomarker expression and drug response .SourceThe OECM-1 human oral cavity squamous cell carcinoma cell line was derived from surgical resection of a primary tumor of a Taiwanese male patient .