Learn More
Braintree Scientific OEM Network Cable for linking (2) OEM Pumps
SDP
Catalog No. 501955430
Encompass
Power supply and secondary cable only. For use when networking two or more OEM BS-500 syringe pumps together. Must use the secondary cable in order to complete this application. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5430 Supplier Braintree Scientific Supplier No. OEMNET
Power supply and secondary cable only. For use when networking two or more OEM BS-500 syringe pumps together. Must use the secondary cable in order to complete this application. **Sold for research purposes only and not for human use**