Learn More
Medchemexpress LLC Oenothein B | 104987-36-2 | 5 MG
SDP
Catalog No. 5000418601
Encompass_Preferred
Oenothein B is a dimeric macrocyclic ellagitannin with broad pharmacological activities, including antioxidant, anti-inflammatory, antifungal, anti-HCV, and antitumor properties. It is also a potent and specific inhibitor of poly(ADP-ribose) glycohydrolase.
  • Dimeric macrocyclic ellagitannin
  • Possesses antioxidant, anti-inflammatory, antifungal, anti-HCV, and antitumor properties
  • Potent and specific inhibitor of poly(ADP-ribose) glycohydrolase
  • Inhibits proliferation of A549 cells by inducing apoptosis and arresting cells at G1 stage
  • Increases intracellular reactive oxygen species (ROS) and upregulates intracellular apoptotic triggers
  • Suppresses lipopolysaccharide (LPS)-induced neuroinflammation in mouse brain
Catalog No. 50-004-18601 Supplier Medchemexpress LLC Supplier No. HYN77655MG
Oenothein B is a dimeric macrocyclic ellagitannin with broad pharmacological activities, including antioxidant, anti-inflammatory, antifungal, anti-HCV, and antitumor properties. It is also a potent and specific inhibitor of poly(ADP-ribose) glycohydrolase.
  • Dimeric macrocyclic ellagitannin
  • Possesses antioxidant, anti-inflammatory, antifungal, anti-HCV, and antitumor properties
  • Potent and specific inhibitor of poly(ADP-ribose) glycohydrolase
  • Inhibits proliferation of A549 cells by inducing apoptosis and arresting cells at G1 stage
  • Increases intracellular reactive oxygen species (ROS) and upregulates intracellular apoptotic triggers
  • Suppresses lipopolysaccharide (LPS)-induced neuroinflammation in mouse brain