Learn More
Medchemexpress LLC OF-02 | 1883431-67-1 | ≥98.0% | C84H152N4O6 | 25 MG
SDP
Catalog No. 5000354097
Encompass_Preferred
OF-02 is an ionizable lipid essential for mRNA delivery, serving as a key component of lipid nanoparticles (LNPs). It becomes protonated in acidic endosomal environments, which aids in the escape of mRNA and effective protein expression in target cells. The lipid's pH-responsive membrane fusion properties allow it to complex with mRNA, disrupt endosomal membrane structures, and facilitate mRNA release into the cytoplasm.
  • Ionizable lipid for mRNA delivery
  • Key component of lipid nanoparticles (LNPs)
  • Promotes endosomal escape of mRNA
  • Induces protein expression in target cells
  • Utilizes pH-responsive membrane fusion properties
  • Enables mRNA release into the cytoplasm
  • Used in mRNA vaccine development
  • Used in gene editing applications
  • Used in protein replacement therapy
Catalog No. 50-003-54097 Supplier Medchemexpress LLC Supplier No. 25MGHY145795
OF-02 is an ionizable lipid essential for mRNA delivery, serving as a key component of lipid nanoparticles (LNPs). It becomes protonated in acidic endosomal environments, which aids in the escape of mRNA and effective protein expression in target cells. The lipid's pH-responsive membrane fusion properties allow it to complex with mRNA, disrupt endosomal membrane structures, and facilitate mRNA release into the cytoplasm.
  • Ionizable lipid for mRNA delivery
  • Key component of lipid nanoparticles (LNPs)
  • Promotes endosomal escape of mRNA
  • Induces protein expression in target cells
  • Utilizes pH-responsive membrane fusion properties
  • Enables mRNA release into the cytoplasm
  • Used in mRNA vaccine development
  • Used in gene editing applications
  • Used in protein replacement therapy