Learn More
Cayman Chemical OF-C4-Deg-Lin, Purity: ≥90%, CAS: 1853203-01-6, 10 mg
SDP
Catalog No. 502916737
Encompass_Preferred
An ionizable cationic lipid; has been used in the generation of LNPs for the delivery of siRNA and mRNA in vitro and in vivo; LNPs containing OF-C4-Deg-Lin and encapsulating an mRNA reporter selectively accumulate in mouse spleen after intravenous administration
Catalog No. 50-291-6737 Supplier Cayman Chemical Supplier No. 3785610 MG
An ionizable cationic lipid; has been used in the generation of LNPs for the delivery of siRNA and mRNA in vitro and in vivo; LNPs containing OF-C4-Deg-Lin and encapsulating an mRNA reporter selectively accumulate in mouse spleen after intravenous administration