Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER WATER STANDARD, 10.0 mg/g | 10 X 5 ML | GFS CHEMICALS, INC. Item #: 99881 | Bulk #: 2303
SDP
Catalog No. NC9740802
Encompass_Preferred
WATERMARK KARL FISCHER WATER STANDARD, 10.0 mg/g | For standardization of volumetric titrants. : NIST Traceable, Water 10.0 mg/g +/- 0.10mg. 9.900 - 10.100
Catalog No. NC9740802 Supplier GFS Chemicals Inc Supplier No. 99881
WATERMARK KARL FISCHER WATER STANDARD, 10.0 mg/g | For standardization of volumetric titrants. : NIST Traceable, Water 10.0 mg/g +/- 0.10mg. 9.900 - 10.100