Learn More
Thermo Scientific Thermostat Assembly Off-Line Source Head for Thermo Scientific ES071 AIG Parts & Accessories [1 pc]
Catalog No. NC0222578 Shop All Thermo Scientific Products
Encompass
Temperature control thermostat with bimetallic strip sensor allows user to adjust activation temperature to maintain constant heat in electric kettles and other small kitchen appliances by cutting power when preset temperature is reached.
Catalog No. NC0222578 Supplier Thermo Scientific Supplier No. ES259
Temperature control thermostat with bimetallic strip sensor allows user to adjust activation temperature to maintain constant heat in electric kettles and other small kitchen appliances by cutting power when preset temperature is reached.