Learn More
Med Vet International Offset Molar Float, 13.5in, J0496A, Each
Catalog No. 5000584847
Encompass_Preferred
These floats are a quality crafted stainless steel float line with many new configurations to employ the newest techniques in equine dental care.Flat shaft provides visualization for blade orientationLightweight and precision balancedFoam non-slip hand grip, colored coded endLarge Allen head set screwFits standard flat float bladesCombined with (JOR57) hardened carbide float blade makes for the finest floating equipment availableBlade not included
Catalog No. 50-005-84847 Supplier Med Vet International Supplier No. JOR496A
These floats are a quality crafted stainless steel float line with many new configurations to employ the newest techniques in equine dental care.Flat shaft provides visualization for blade orientationLightweight and precision balancedFoam non-slip hand grip, colored coded endLarge Allen head set screwFits standard flat float bladesCombined with (JOR57) hardened carbide float blade makes for the finest floating equipment availableBlade not included