Learn More
Foxx Life Sciences EZBio Stainless Steel Offset Spoon Polished with Steel Handle, 304 SS, 300mm, 1/EA
Catalog No. NC1795112
Encompass_Preferred
Foxx Life Sciences Stainless Steel Offset Spoon-Spatulas are designed for ease of use, flowability, and versatility. This double-sided, multifunctional device features an offset spoon on one side and a spatula on the other for an easy pour into containers. These ideal products make it possible to support numerous processes, such as formulation work, measuring, sample gathering, powder prep, granulate work, with a variety of media including pastes and liquids. Manufactured with a single piece of high-quality 304 Stainless Steel, they are free of edges, crevices, or undercuts, to eliminate the possibility of cross contamination, product residue, or cleaning agent deposits. They are perfect for uses in chemistry, pharmaceuticals, cosmetics, and food. These double-sided spoons are available in range of sizes, from 150mm to 500mm.304 Stainless Steel High-gloss polishedDouble-sided functionalityEasy to clean hygienic design
Catalog No. NC1795112 Supplier Foxx Life Sciences Supplier No. 25C3608FLS
Foxx Life Sciences Stainless Steel Offset Spoon-Spatulas are designed for ease of use, flowability, and versatility. This double-sided, multifunctional device features an offset spoon on one side and a spatula on the other for an easy pour into containers. These ideal products make it possible to support numerous processes, such as formulation work, measuring, sample gathering, powder prep, granulate work, with a variety of media including pastes and liquids. Manufactured with a single piece of high-quality 304 Stainless Steel, they are free of edges, crevices, or undercuts, to eliminate the possibility of cross contamination, product residue, or cleaning agent deposits. They are perfect for uses in chemistry, pharmaceuticals, cosmetics, and food. These double-sided spoons are available in range of sizes, from 150mm to 500mm.304 Stainless Steel High-gloss polishedDouble-sided functionalityEasy to clean hygienic design