Learn More
Medchemexpress LLC HY-B0125 500mg Medchemexpress, Ofloxacin CAS:82419-36-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0125500MG
Encompass_Preferred
Medchemexpress, HY-B0125 500mg Ofloxacin CAS:82419-36-1 Ofloxacin (Hoe-280) is a fluoroquinolone whose primary mechanism of action is inhibition of bacterial DNA gyrase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2371