Learn More
Medchemexpress LLC Ofloxacin | 82419-36-1 | 99.9% | 250 MG
SDP
Catalog No. 5000406820
Encompass_Preferred
Ofloxacin is a fluoroquinolone antibiotic that primarily inhibits bacterial DNA gyrase. It demonstrates inhibitory activity against vaccinia virus (VV) and possesses a broad spectrum of activity against aerobic Gram-negative and Gram-positive bacteria. Unlike many other frontline antibacterial drugs, it specifically targets bacterial DNA synthesis rather than cell wall or protein synthesis.
  • Inhibits bacterial DNA gyrase
  • Exhibits activity against vaccinia virus
  • Offers broad spectrum activity against aerobic Gram-negative and Gram-positive bacteria
  • Targets bacterial DNA synthesis
Catalog No. 50-004-06820 Supplier Medchemexpress LLC Supplier No. 250MGHYB0125R
Add to Cart
Add to Cart
Ofloxacin is a fluoroquinolone antibiotic that primarily inhibits bacterial DNA gyrase. It demonstrates inhibitory activity against vaccinia virus (VV) and possesses a broad spectrum of activity against aerobic Gram-negative and Gram-positive bacteria. Unlike many other frontline antibacterial drugs, it specifically targets bacterial DNA synthesis rather than cell wall or protein synthesis.
  • Inhibits bacterial DNA gyrase
  • Exhibits activity against vaccinia virus
  • Offers broad spectrum activity against aerobic Gram-negative and Gram-positive bacteria
  • Targets bacterial DNA synthesis