Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ofloxacin-d3 | 1173147-91-5 | 364.39 g/mol | C18H17D3FN3O4 | 10 MG
SDP

Catalog No. 5000298857
Encompass_Preferred

Ofloxacin-d3 is the deuterium-labeled analogue of the fluoroquinolone antibiotic ofloxacin, supplied for research and analytical applications such as quantitative mass spectrometry and tracer studies. It is intended for research use only and not for human or clinical use.

  • Deuterium-labeled internal standard for LC-MS quantitation.
  • Chemical formula C18H17D3FN3O4 and molecular weight 364.39 g/mol.
  • CAS number 1173147-91-5 for unambiguous identification.
  • Available in small research pack sizes, including 10 MG.
  • Powder storage: -20°C (stable for up to 3 years); in solvent: short-term storage per supplier guidance.

Catalog No. 50-002-98857 Supplier Medchemexpress LLC Supplier No. HYB0125S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ofloxacin-d3 is the deuterium-labeled analogue of the fluoroquinolone antibiotic ofloxacin, supplied for research and analytical applications such as quantitative mass spectrometry and tracer studies. It is intended for research use only and not for human or clinical use.

  • Deuterium-labeled internal standard for LC-MS quantitation.
  • Chemical formula C18H17D3FN3O4 and molecular weight 364.39 g/mol.
  • CAS number 1173147-91-5 for unambiguous identification.
  • Available in small research pack sizes, including 10 MG.
  • Powder storage: -20°C (stable for up to 3 years); in solvent: short-term storage per supplier guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.