Learn More
Medchemexpress LLC Ofloxacin-d8 | 1219170-21-4 | 99.9% | 1 MG
SDP
Catalog No. 5000450505
Encompass_Preferred
Ofloxacin-d8 (Hoe-280-d8) is the deuterium-labeled Ofloxacin. Ofloxacin (Hoe-280) is a fluoroquinolone that inhibits bacterial DNA gyrase and is for research use only.
  • Deuterium-labeled compound
  • Inhibits bacterial DNA gyrase
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-50505 Supplier Medchemexpress LLC Supplier No. HYB0125S11MG
Ofloxacin-d8 (Hoe-280-d8) is the deuterium-labeled Ofloxacin. Ofloxacin (Hoe-280) is a fluoroquinolone that inhibits bacterial DNA gyrase and is for research use only.
  • Deuterium-labeled compound
  • Inhibits bacterial DNA gyrase
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS