Learn More
Medchemexpress LLC Ofloxacin-d8 | 1219170-21-4 | 99.9% | 5 MG
SDP
Catalog No. 5000450850
Encompass_Preferred
Ofloxacin-d8 (Hoe-280-d8) is the deuterium labeled Ofloxacin. It is a fluoroquinolone that primarily inhibits bacterial DNA gyrase. Intended for research use only, this compound can serve as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. For storage, the powder form should be kept at -20°C for 3 years. If in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Deuterium labeled Ofloxacin
  • Inhibits bacterial DNA gyrase
  • Can be used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for quantitation during drug development
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-50850 Supplier Medchemexpress LLC Supplier No. HYB0125S15MG
Ofloxacin-d8 (Hoe-280-d8) is the deuterium labeled Ofloxacin. It is a fluoroquinolone that primarily inhibits bacterial DNA gyrase. Intended for research use only, this compound can serve as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. For storage, the powder form should be kept at -20°C for 3 years. If in solvent, store at -80°C for 6 months or -20°C for 1 month.
  • Deuterium labeled Ofloxacin
  • Inhibits bacterial DNA gyrase
  • Can be used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for quantitation during drug development
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs