Learn More
Enzo Life Sciences n-Octyl-beta-D-glucopyranoside (analytical grade) (1g). CAS: 29836-26-8
SDP
Supplier:  Enzo Life Sciences ALX500001G001
Encompass_Preferred
Highly purified nonionic detergent that solubilizes membrane-bound proteins in their native state. Alternative name: OG. Purity: ≥99% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water (100mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9334