Learn More
Aviva Systems Biology Corp OGDH Antibody - N-terminal region (ARP58648_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58648_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OGDH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OGDH. Alternate names: AKGDH, E1k, OGDCApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62387