Learn More
Aviva Systems Biology Corp OGDHL Antibody - N-terminal region (ARP57175_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57175_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OGDHL. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human OGDHL. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-286-272