Learn More
ABclonal Technology OGDHL Rabbit pAb
SDP
Supplier:  ABclonal Technology A154751000UL
Encompass_Preferred
The protein encoded by this gene is similar to oxoglutarate dehydrogenase (OGDH) of the OGDH complex, which degrades glucose and glutamate. This gene encodes several isoforms, including some that appear to localize to mitochondria. The encoded protein down-regulates the AKT signaling cascade and can suppress the growth of cervical cancer cells.
Catalog No. 50-001-05955