Learn More
Medchemexpress LLC Ogerin | 1309198-71-7 | 99.9% | 307.35 | 10 MG
SDP
Catalog No. 501968786
Encompass_Preferred
Ogerin is a chemical probe that acts as a selective GPR68 positive allosteric modulator (PAM) and has a moderate antagonistic effect on A2A. It inhibits the fear conditioning reflex in mice and prevents TGF-β-induced myofibroblast differentiation of fibroblasts. It is useful in studies of fibrotic diseases and neurological disorders.
  • Acts as a selective GPR68 positive allosteric modulator (PAM)
  • Exhibits moderate antagonistic effect on A2A
  • Inhibits fear conditioning reflex in mice
  • Prevents TGF-β-induced myofibroblast differentiation of fibroblasts
  • Useful for studying fibrotic diseases and neurological disorders
Catalog No. 50-196-8786 Supplier Medchemexpress LLC Supplier No. HY11027910MG
Ogerin is a chemical probe that acts as a selective GPR68 positive allosteric modulator (PAM) and has a moderate antagonistic effect on A2A. It inhibits the fear conditioning reflex in mice and prevents TGF-β-induced myofibroblast differentiation of fibroblasts. It is useful in studies of fibrotic diseases and neurological disorders.
  • Acts as a selective GPR68 positive allosteric modulator (PAM)
  • Exhibits moderate antagonistic effect on A2A
  • Inhibits fear conditioning reflex in mice
  • Prevents TGF-β-induced myofibroblast differentiation of fibroblasts
  • Useful for studying fibrotic diseases and neurological disorders