Learn More
Aviva Systems Biology Corp OGFOD1 Antibody - middle region (ARP48825_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48825_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OGFOD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OGFOD1. Alternate names: FLJ10826, KIAA1612, TPA1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53563