Learn More
Medchemexpress LLC OGG1 Human N-His 10ug
SDP
Catalog No. 5000182938
Encompass_Preferred
The OGG1 protein is a DNA repair enzyme specifically designed to cleave DNA at the 8-oxoG residue and plays a key role in maintaining genome integrity It demonstrates the ability to excise damaged bases such as 7 8-dihydro-8-oxoguanine and 2 6-diamino-4-hydroxy-5-N-methylformamidopyrimidine from the DNA structure (FAPY) OGG1 Protein Human (N-His) is the recombinant human-derived OGG1 protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-82938 Supplier Medchemexpress LLC Supplier No. HYP72338A10UG
The OGG1 protein is a DNA repair enzyme specifically designed to cleave DNA at the 8-oxoG residue and plays a key role in maintaining genome integrity It demonstrates the ability to excise damaged bases such as 7 8-dihydro-8-oxoguanine and 2 6-diamino-4-hydroxy-5-N-methylformamidopyrimidine from the DNA structure (FAPY) OGG1 Protein Human (N-His) is the recombinant human-derived OGG1 protein expressed by E coli with N-6 His labeled tag