Learn More
TARGETMOL CHEMICALS INC OGLEMILAST 5MG
SDP
Catalog No. 502581181
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Oglemilast (GRC 3886) suppresses pulmonary cell infiltration including eosinophilia and neutrophilia in vitro and in vivo. Oglemilast is an effective and orally active phosphodiesterase-4 (PDE4) inhibitor (IC50 0.5 nM for PDE4D3). Oglemilast has the potential for inflammatory airway diseases. purity: 98%
Catalog No. 50-258-1181 Supplier TARGETMOL CHEMICALS INC Supplier No. T163795MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Oglemilast (GRC 3886) suppresses pulmonary cell infiltration including eosinophilia and neutrophilia in vitro and in vivo. Oglemilast is an effective and orally active phosphodiesterase-4 (PDE4) inhibitor (IC50 0.5 nM for PDE4D3). Oglemilast has the potential for inflammatory airway diseases. purity: 98%