Learn More
GENOVIS INC Lyophilized OglyZOR, 2000 units
Catalog No. NC1337652
Encompass
OglyZOR enzyme for deglycosylation of 2 mg glycoprotein, including one vial of SialEXO for sialic acid removal.
This item is not returnable. View return policy
Catalog No. NC1337652 Supplier GENOVIS INC Supplier No. G2OG1020
OglyZOR enzyme for deglycosylation of 2 mg glycoprotein, including one vial of SialEXO for sialic acid removal.