Learn More
Lonza Walkersville OGM Osteoblast Growth Medium BulletKit
Catalog No. NC9732095
Encompass
Clonetics OGM Osteoblast Growth Medium is specially formulated for the growth and expansion of human osteoblasts. The kit contains 10% serum. The kit is conveniently packaged to include basal medium with growth supplements for complete culture of osteoblasts.OGM Differentiation Supplements are also available and sold separately (catalog number CC-4194). These are recommended for use with differentiation assays. Applications:In vitro bone modelsMineralizationOsteoporosisGene expressionHormone treatmentsOsteopeniaPaget's DiseaseContent:1 x Bottle Basal Medium 500 mL1 x Bottle FBS 50.00 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Blue Cap Vial with Ascorbic Acid 0.50 mL
Catalog No. NC9732095 Supplier Lonza Walkersville Supplier No. CC3207
Add to Cart
Add to Cart
Clonetics OGM Osteoblast Growth Medium is specially formulated for the growth and expansion of human osteoblasts. The kit contains 10% serum. The kit is conveniently packaged to include basal medium with growth supplements for complete culture of osteoblasts.OGM Differentiation Supplements are also available and sold separately (catalog number CC-4194). These are recommended for use with differentiation assays. Applications:In vitro bone modelsMineralizationOsteoporosisGene expressionHormone treatmentsOsteopeniaPaget's DiseaseContent:1 x Bottle Basal Medium 500 mL1 x Bottle FBS 50.00 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Blue Cap Vial with Ascorbic Acid 0.50 mL