Learn More
Lonza Walkersville OGM Osteoblast Growth Med Sin
Supplier:  Lonza Walkersville CC4193
Encompass
OGM SingleQuot Kit are components sold separately and does not include the basal medium These are part of the OGM Osteoblast Growth Medium BulletKit (Catalog Number CC-3207) The full OGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3207 Clonetics OGM Osteoblast Growth Medium is specially formulated for the growth and expansion of human osteoblasts The kit contains 10% serum OGM Differentiation Supplements are also included and are recommended for use with differentiation assays The kit is conveniently packaged to include basal medium with growth and differentiation supplements for complete culture of osteoblasts
Catalog No. 50-004-96379