Learn More
TARGETMOL CHEMICALS INC OGT inhibitor 4a 1MG
SDP
Catalog No. 502877036
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. OGT inhibitor 4a is an O-linked n-acetylglucosaminyltransferase (OGT) inhibitor with anticancer activity for the study of autoimmune and neurological disorders. Purity 99.08%
Catalog No. 50-287-7036 Supplier TARGETMOL CHEMICALS INC Supplier No. T884381MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. OGT inhibitor 4a is an O-linked n-acetylglucosaminyltransferase (OGT) inhibitor with anticancer activity for the study of autoimmune and neurological disorders. Purity 99.08%