Learn More
Apexbio Technology LLC OH-CITRIC ACID TRIK-500mg
SDP
Catalog No. 5000897553
Encompass_Preferred
Hydroxycitric acid tripotassium hydrate (CAS No 6100-05-6) is a compound commonly used in biomedical research especially in studies related to metabolic regulation and weight management Originating from certain tropical plants it primarily acts by inhibiting the citrate lyase enzyme a key catalyst in the conversion of excess carbohydrates into fat Its mechanism of action influences various pathways involved in lipid synthesis and energy balance In vitro it demonstrates activity in nanomolar to low micromolar ranges making it suitable for precise biochemical assays This compound is frequently utilized in cell models and animal studies to explore its potential therapeutic effects and efficacy Moreover it is a candidate in drug screening programs focusing on metabolic disorders Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97553 Supplier Apexbio Technology LLC Supplier No. C6518500
Hydroxycitric acid tripotassium hydrate (CAS No 6100-05-6) is a compound commonly used in biomedical research especially in studies related to metabolic regulation and weight management Originating from certain tropical plants it primarily acts by inhibiting the citrate lyase enzyme a key catalyst in the conversion of excess carbohydrates into fat Its mechanism of action influences various pathways involved in lipid synthesis and energy balance In vitro it demonstrates activity in nanomolar to low micromolar ranges making it suitable for precise biochemical assays This compound is frequently utilized in cell models and animal studies to explore its potential therapeutic effects and efficacy Moreover it is a candidate in drug screening programs focusing on metabolic disorders Experimental concentrations typically depend on the specific research design