Learn More
Medchemexpress LLC Oh-glu-val-cit-pab-mmae | 1895916-23-0 | MFCD34676998 | 99.0% | 1237.53 | C63H100N10O15 | 5 MG
SDP
Catalog No. 5000203595
Encompass_Preferred
A cleavable antibody-drug conjugate (ADC) reagent combining an OH-Glu-Val-Cit-PAB cleavable linker with the cytotoxic payload monomethyl auristatin E (MMAE), supplied for research use in ADC synthesis.
  • Cleavable linker conjugated to a tubulin inhibitor for ADC construction.
  • High purity suitable for research applications.
  • Supplied as a 5 mg unit; larger sizes available on request.
  • Store sealed at -20°C, protected from moisture and light; in solvent store at -80°C for up to 6 months.
  • Includes CAS 1895916-23-0 and molecular weight 1237.53 for reference.
Catalog No. 50-002-03595 Supplier Medchemexpress LLC Supplier No. HY1482455MG
A cleavable antibody-drug conjugate (ADC) reagent combining an OH-Glu-Val-Cit-PAB cleavable linker with the cytotoxic payload monomethyl auristatin E (MMAE), supplied for research use in ADC synthesis.
  • Cleavable linker conjugated to a tubulin inhibitor for ADC construction.
  • High purity suitable for research applications.
  • Supplied as a 5 mg unit; larger sizes available on request.
  • Store sealed at -20°C, protected from moisture and light; in solvent store at -80°C for up to 6 months.
  • Includes CAS 1895916-23-0 and molecular weight 1237.53 for reference.