Learn More
Research Products International Corp OHAUS Adventurer Analytical Balance, AX224/E
SDP
Catalog No. 502114193
Encompass
Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities. With a color touchscreen, GLP/GMP compliance capabilities, two USB ports, and much more, Adventurer is the most complete balance in its class.
Unit is not returnable without original packaging box and material. There are no exceptions.
Catalog No. 50-211-4193 Supplier Research Products International Corp Supplier No. OH30100603
Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities. With a color touchscreen, GLP/GMP compliance capabilities, two USB ports, and much more, Adventurer is the most complete balance in its class.
Unit is not returnable without original packaging box and material. There are no exceptions.