Learn More
Research Products International Corp OHAUS Adventurer Precision Balances, Model AX4202/E
SDP
Catalog No. 502114121
Encompass
Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities. With a color touchscreen, GLP/GMP compliance capabilities, two USB ports, and much more, Adventurer is the most complete balance in its class.
Adventurer balances come with a color touchscreen, an icon-based user interface, and an ergonomic design, making them easy to configure and use.
Features such as specialized weighing modes, multiple connectivity options, and AutoCal‚Ñ¢ provide versatility and flexibility for a variety of applications.
Durable construction, large weighing surfaces, a space-saving draftshield design, and full housing in-use cover allow for use in lab, education and industrial environments.
Unit is not returnable without original packaging box and material. There are no exceptions.
Catalog No. 50-211-4121 Supplier Research Products International Corp Supplier No. OH30100616
Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities. With a color touchscreen, GLP/GMP compliance capabilities, two USB ports, and much more, Adventurer is the most complete balance in its class.
Adventurer balances come with a color touchscreen, an icon-based user interface, and an ergonomic design, making them easy to configure and use.
Features such as specialized weighing modes, multiple connectivity options, and AutoCal‚Ñ¢ provide versatility and flexibility for a variety of applications.
Durable construction, large weighing surfaces, a space-saving draftshield design, and full housing in-use cover allow for use in lab, education and industrial environments.
Unit is not returnable without original packaging box and material. There are no exceptions.