Learn More
OHAUS™ Adventurer™ Analytical Balances
Catalog No. 0192830 Shop All OHAUS Products
Change view
Click to view available options
Model:
AR1140-2E0
Readability:
0.1 mg
Stabilization Time:
3 sec.
Catalog No. Model Readability Stabilization Time
01-928-30 AR1140-2E0 0.1 mg 3 sec.
1 options
Catalog No. 01-928-30 Supplier OHAUS™ Supplier No. RSD/80103738
Analytical precision in an economical balance; Capacities to 210g; Multiple weighing units; Easy to use.
The perfect choice for budget-conscious laboratories. Adventurer analytical balances offer precision weighing, dependability and ease of use at an economical price. Three models with capacities of 65, 110, and 210g, all with readability to 0.1mg. Standard features include stability indicator, error indicator, three-door draft shield, RS-232-C interface, weigh-below hook, integral security bracket, 15 weighing units and parts counting.
Simple operation
  • Three-button operation with separate PRINT button
  • Large left and right TARE buttons for convenient use with either hand
  • Simple-to-use set-up menus allow you to select weighing units, calibrate the balance, perform parts counting and set print parameters
  • An audible signal lets you know that the key you pressed is working correctly
  • Results are displayed on a large, easy-to-read LCD
Ergonomic design
  • Angled keypad enhances viewing
  • Contoured base makes it easy to pick up and move balance
  • Ergonomically designed leveling system with level bubble and adjustable feet assures that the balance remains level
Easy calibration
  • To access the calibration menu, place calibration weight on pan, then press a button
Multiple weighing units and parts counting handle all basic weighing needs
  • Sixteen user-selectable weighing units include: grams, kilograms, milligrams, carats, Newtons, pounds, ounces, troy ounces, grains, pennyweight, mommes, mesghals, three taels (Hong Kong, Singapore and Taiwan), and ticals
  • Balance counts parts based on the weight of a reference sample of 5, 10, 20, 50 or 100 parts
Built-in RS-232-C interface
  • Standard RS-232-C interface allows connection to computers, printers and other peripheral devices
  • Operates bidirectionally at user-selectable transfer rates of 600, 1200, 2400, 4800 or 9600bps
  • Manually print only stable or all data, or set balance to automatically print stable data when display changes by at least five counts
Virtually maintenance-free
  • Solid die-cast metal base stands up to heavy laboratory or classroom use
  • Weighing pan is made of chemical-resistant stainless steel
  • Sealed keypad and molded spill ring protect balance for years of uninterrupted use
  • Durable, three-door draft shield protects weighing chamber and allows convenient and stable weighing
  • Balances operate on supplied AC adapter
Warranty and Services
Two-year warranty.
TRUSTED_SUSTAINABILITY
Specifications
Readability 0.1 mg
Model AR1140-2E0
Stabilization Time 3 sec.