Learn More
Research Products International Corp OHAUS 2 Block Dry Block Heater, Digital, HB2DG
SDP
Catalog No. 502229947
Encompass
Versatile Dry Block Heaters are ideal for applications that require temperature stability. All units hold 2 optional interchangeable modular blocks with over 40 choices. Blocks are easy to use, no tools required. Close tube-to-block contact enables maximum heat retention, resulting in efficient heating. Digital models offer exceptional temperature uniformity and stability for repeatable results, while non-digital models are an economical choice.
Three models to customize your application. All models offer exceptional temperature uniformity with outstanding temperature stability.
Avoid contamination to ensure repeatable, reliable results. The Heatblock with Heated Lid features a second heater in the lid to minimize condensation to maintain your sample integrity.
Ensure temperature accuracy.
Accepts block with dimensions: 3.75 x 3 x 2 H inches
Catalog No. 50-222-9947 Supplier Research Products International Corp Supplier No. OH30392080
Versatile Dry Block Heaters are ideal for applications that require temperature stability. All units hold 2 optional interchangeable modular blocks with over 40 choices. Blocks are easy to use, no tools required. Close tube-to-block contact enables maximum heat retention, resulting in efficient heating. Digital models offer exceptional temperature uniformity and stability for repeatable results, while non-digital models are an economical choice.
Three models to customize your application. All models offer exceptional temperature uniformity with outstanding temperature stability.
Avoid contamination to ensure repeatable, reliable results. The Heatblock with Heated Lid features a second heater in the lid to minimize condensation to maintain your sample integrity.
Ensure temperature accuracy.
Accepts block with dimensions: 3.75 x 3 x 2 H inches